Learn More
Biomatik Corporation Human PRKAR1A (Protein kinase cAMP-dependent type I regulatory subunit alpha) ELISA Kit
Supplier:  Biomatik Corporation EKF60146
Encompass_Preferred
Human PRKAR1A (Protein kinase cAMP-dependent type I regulatory subunit alpha) ELISA Kit for detecting PRKAR1A in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P10644. Target Synonyms: CAR; CNC; CNC1; PKR1; PPNAD1; PRKAR1; PRKAR1A; Tissue specific extinguisher 1; TSE1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6451