Learn More
Biomatik Corporation Human MLK4 (Mitogen-activated protein kinase kinase kinase 21) ELISA Kit
Supplier:  Biomatik Corporation EKF60173
Encompass_Preferred
Human MLK4 (Mitogen-activated protein kinase kinase kinase 21) ELISA Kit for detecting MLK4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q5TCX8. Target Synonyms: KIAA1804; mixed lineage kinase 4; MLK4; RP5 862P8.2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6454