Learn More
Biomatik Corporation Porcine GPX1 (Glutathione peroxidase 1) ELISA Kit
Supplier:  Biomatik Corporation EKF60228
Encompass_Preferred
Porcine GPX1 (Glutathione peroxidase 1) ELISA Kit for detecting GPX1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8MJ14. Target Synonyms: Gpx1; CGPx; GPX1; GSHPX1; Cellular glutathione peroxidase; EC 1.11.1; EC 1.11.1.9; glutathione peroxidase 1; GPx-1; GSHPX1; GSHPx-1; MGC14399; MGC88245. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6456