Learn More
Biomatik Corporation Rat MMP-7 (Matrix Metalloproteinase-7) ELISA Kit
Supplier:  Biomatik Corporation EKF60385
Encompass_Preferred
Rat MMP-7 (Matrix Metalloproteinase-7) ELISA Kit for detecting MMP-7 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P50280. Target Synonyms: MMP-7; Matrix Metalloproteinase 7; Matrilysin; matrilysin; matrin; Matrin; MPSL1; PUMP1; PUMP-1; Pump-1 protease; uterine matrilysin; Uterine metalloproteinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6461