Learn More
Biomatik Corporation Human FUT3 (Galactoside 3 (4)-L-fucosyltransferase) ELISA Kit
Supplier:  Biomatik Corporation EKF60430
Encompass_Preferred
Human FUT3 (Galactoside 3 (4)-L-fucosyltransferase) ELISA Kit for detecting FUT3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P21217. Target Synonyms: FUT3; CD174; FT3B; FucT-III; Les; Lewis FT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6478