Learn More
Biomatik Corporation Rat CA125 (CarbohydRate Antigen 125) ELISA Kit
Supplier:  Biomatik Corporation EKF60440
Encompass_Preferred
Rat CA125 (CarbohydRate Antigen 125) ELISA Kit for detecting CA125 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.813-500iu/ml. Sensitivity: 4.688iu/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: D3ZM49. Target Synonyms: CA125; CA-125; MUC16; Mucin 16. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6486