Learn More
Biomatik Corporation Human TSH (Thyroid Stimulating Hormone) ELISA Kit
Supplier:  Biomatik Corporation EKF60486
Encompass_Preferred
Human TSH (Thyroid Stimulating Hormone) ELISA Kit for detecting TSH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.0156-1uiu/ml. Sensitivity: 0.0094uiu/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01222; P01215. Target Synonyms: TSH; Thyroid Stimulating Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6513