Learn More
Biomatik Corporation Human TIMP-3 (Metalloproteinase inhibitor 3) ELISA Kit
Supplier:  Biomatik Corporation EKF60488
Encompass_Preferred
Human TIMP-3 (Metalloproteinase inhibitor 3) ELISA Kit for detecting TIMP-3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P35625. Target Synonyms: TIMP-3; MIG-5 protein; Protein MIG-5; SFD; TIMP metallopeptidase inhibitor 3; tissue inhibitor of metalloproteinase 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6515