Learn More
Biomatik Corporation Human IL5RA (Interleukin-5 receptor) ELISA Kit
Supplier:  Biomatik Corporation EKF60492
Encompass_Preferred
Human IL5RA (Interleukin-5 receptor) ELISA Kit for detecting IL5RA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q01344. Target Synonyms: IL5RA; CD125; IL5R; CD125 antigen; IL-5RA; CDw125; HSIL5R3; IL-5 receptor subunit alpha; IL-5R subunit alpha; IL-5R-alpha; interleukin 5 receptor type 3; interleukin 5 receptor; alpha; interleukin-5 receptor alpha chain; interleukin-5 receptor alpha subunit; interleukin-5 receptor subunit alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6519