Learn More
Biomatik Corporation Human F12 (Coagulation factor XII) ELISA Kit
Supplier:  Biomatik Corporation EKF60494
Encompass_Preferred
Human F12 (Coagulation factor XII) ELISA Kit for detecting F12 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P00748. Target Synonyms: F12; Coagulation factor XII; HAEX; Hageman factor; HAE3; HAF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6521