Learn More
Biomatik Corporation Mouse MMP-12 (Matrix Metalloproteinase 12) ELISA Kit
Supplier:  Biomatik Corporation EKF60507
Encompass_Preferred
Mouse MMP-12 (Matrix Metalloproteinase 12) ELISA Kit for detecting MMP-12 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P34960. Target Synonyms: MMP-12; MMP12; HME; ME; MME; Macrophage Metalloelastase; Macrophage Elastase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6533