Learn More
Biomatik Corporation Mouse MMP-10 (Matrix Metalloproteinase 10) ELISA Kit
Supplier:  Biomatik Corporation EKF60522
Encompass_Preferred
Mouse MMP-10 (Matrix Metalloproteinase 10) ELISA Kit for detecting MMP-10 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O55123. Target Synonyms: MMP-10; SL-2; STMY2; Stromelysin-2; Transin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6547