Learn More
Biomatik Corporation Mouse ECF/CCL11 (Eosinophil Chemotactic Factor) ELISA Kit
Supplier:  Biomatik Corporation EKF60532
Encompass_Preferred
Mouse ECF/CCL11 (Eosinophil Chemotactic Factor) ELISA Kit for detecting ECF; CCL11 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P48298. Target Synonyms: CCL11; ECF; Eotaxin; chemokine ligand 11; Eotaxin-1; Eosinophil chemotactic protein; SCYA11Small-inducible cytokine A11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6556