Learn More
Biomatik Corporation Mouse GROγ (Growth Regulated Oncogene Gamma) ELISA Kit
Supplier:  Biomatik Corporation EKF60537
Encompass_Preferred
Mouse GROγ (Growth Regulated Oncogene Gamma) ELISA Kit for detecting GROγ in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q6W5C0. Target Synonyms: GROγ; CXCL3; CINC-2; DCIP-1; GRO gamma; Dcip1; CINC-2b; GRO3; GRO3 oncogene; GROg; GROG; GRO-gamma; MIP2B; MIP-2b; MIP2-beta; SCYB3; Gm1960; chemokine; C-X-C motif ligand 3; C-X-C motif chemokine 3; GRO-gamma; 1-73; Growth-regulated protein gamma; Macrophage inflammatory protein 2-beta; melanoma growth stimulatory activity gamma; MGSA gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6560