Learn More
Biomatik Corporation Mouse HSP-27 (heat shock protein 27) ELISA Kit
Supplier:  Biomatik Corporation EKF60539
Encompass_Preferred
Mouse HSP-27 (heat shock protein 27) ELISA Kit for detecting HSP-27 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P14602. Target Synonyms: HSP27; Heat Shock Protein 27; HSP-27; HSPB1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6562