Learn More
Biomatik Corporation Rat SELE (E-Selectin) ELISA Kit
Supplier:  Biomatik Corporation EKF60552
Encompass_Preferred
Rat SELE (E-Selectin) ELISA Kit for detecting SELE in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P98105. Target Synonyms: E-Selectin; SELE; CD62E; CD62E; ELAM1; LECAM2; SELE; CD62 antigen-like family member E; CD62E; CD62E antigen; ELAM; ELAM-14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6575