Learn More
Biomatik Corporation Rat 5-LO (Arachidonate 5-Lipoxygenase) ELISA Kit
Supplier:  Biomatik Corporation EKF60566
Encompass_Preferred
Rat 5-LO (Arachidonate 5-Lipoxygenase) ELISA Kit for detecting 5-LO in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P12527. Target Synonyms: 5-LO; Alox5; 5-LO; 5LPG; LOG5; LOX5A; 5-lipoxygenase; 5-LOX; arachidonic 5-lipoxygenase alpha-10 isoform; arachidonic 5-lipoxygenase delta-10-13 isoform; arachidonic 5-lipoxygenase delta-13 isoform; arachidonic 5-lipoxygenase delta-p10 isoform; arachidonic acid 5-lipoxygenase; leukotriene A4 synthase; LOG5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6589