Learn More
Biomatik Corporation Human FGα (Fibrinogen Alpha) ELISA Kit
Supplier:  Biomatik Corporation EKF60573
Encompass_Preferred
Human FGα (Fibrinogen Alpha) ELISA Kit for detecting FGα in plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.469-30ng/ml. Sensitivity: 0.281ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02671. Target Synonyms: FGα; Factor I; FGA; FGB; Fib2; Fib2; fibrinogen alpha chain; fibrinogen; A alpha polypeptide; MGC119422; MGC119423; MGC119425. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6596