Learn More
Biomatik Corporation Human FBXO32 (F-box only protein 32) ELISA Kit
Supplier:  Biomatik Corporation EKF60579
Encompass_Preferred
Human FBXO32 (F-box only protein 32) ELISA Kit for detecting FBXO32 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q969P5. Target Synonyms: FLJ32424; Fbx32; MAFbx; MGC33610; F-box only protein 32; atrogin 1; muscle atrophy F-box protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6602