Learn More
Biomatik Corporation Rat Cyt-C (Cytochrome C) ELISA Kit
Supplier:  Biomatik Corporation EKF60586
Encompass_Preferred
Rat Cyt-C (Cytochrome C) ELISA Kit for detecting Cyt-C in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.391-25ng/ml. Sensitivity: 0.234ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P62898. Target Synonyms: Cyt-C; CYCS; CYCHCS; cytochrome c; cytochrome c; somatic; THC4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6609