Learn More
Biomatik Corporation Rat Maoa (Monoamine Oxidase Type A) ELISA Kit
Supplier:  Biomatik Corporation EKF60592
Encompass_Preferred
Rat Maoa (Monoamine Oxidase Type A) ELISA Kit for detecting Maoa in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P21396. Target Synonyms: Maoa; MAO-A; amine oxidase [flavin-containing] A; monoamine oxidase A; Monoamine oxidase type A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6615