Learn More
Biomatik Corporation Rat Tg (Thyroglobulin) ELISA Kit
Supplier:  Biomatik Corporation EKF60614
Encompass_Preferred
Rat Tg (Thyroglobulin) ELISA Kit for detecting Tg in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P06882. Target Synonyms: Tg. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6637