Learn More
Biomatik Corporation Human IFNGR1 (Interferon gamma receptor 1) ELISA Kit
Supplier:  Biomatik Corporation EKF60635
Encompass_Preferred
Human IFNGR1 (Interferon gamma receptor 1) ELISA Kit for detecting IFNGR1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P15260. Target Synonyms: IFNGR1; CD119; AVP; type 2; CDw119; IFNGR; CD119 antigen; IFN-gamma receptor 1; IFN-gamma-R1; immune interferon receptor 1; interferon gamma receptor 1; interferon-gamma receptor alpha chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6656