Learn More
Biomatik Corporation Human SESN3 (Sestrin-3) ELISA Kit
Supplier:  Biomatik Corporation EKF60637
Encompass_Preferred
Human SESN3 (Sestrin-3) ELISA Kit for detecting SESN3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P58005. Target Synonyms: SESN3; Sestrin-3; SEST3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6658