Learn More
Biomatik Corporation Rat Aldh2 (Aldehyde dehydrogenase, mitochondrial) ELISA Kit
Supplier:  Biomatik Corporation EKF60650
Encompass_Preferred
Rat Aldh2 (Aldehyde dehydrogenase, mitochondrial) ELISA Kit for detecting Aldh2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P11884. Target Synonyms: ALDH2; Aldehyde dehydrogenase; mitochondrial; ALDHI; ALDH class 2; ALDH-E2; ALDM; Aldehyde Dehydrogenase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6667