Learn More
Biomatik Corporation Mouse Irf5 (Interferon regulatory factor 5) ELISA Kit
Supplier:  Biomatik Corporation EKF60659
Encompass_Preferred
Mouse Irf5 (Interferon regulatory factor 5) ELISA Kit for detecting Irf5 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P56477. Target Synonyms: Irf5; IBD14; interferon regulatory factor 5; IRF-5; SLEB10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6675