Learn More
Biomatik Corporation Mouse Romo1 (Reactive oxygen species modulator 1) ELISA Kit
Supplier:  Biomatik Corporation EKF60663
Encompass_Preferred
Mouse Romo1 (Reactive oxygen species modulator 1) ELISA Kit for detecting Romo1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P60603. Target Synonyms: Romo1; Reactive oxygen species modulator 1; ROS modulator 1; Protein MGR2 homolog; Mitochondrial targeting GxxxG motif protein; MTGM; Protein MGR2 homolog; Glyrichin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6678