Learn More
Biomatik Corporation Human EphA2 (Tyrosine protein kinase receptor A2) ELISA Kit
Supplier:  Biomatik Corporation EKF60665
Encompass_Preferred
Human EphA2 (Tyrosine protein kinase receptor A2) ELISA Kit for detecting EphA2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P29317. Target Synonyms: EphA2; Eck; Myk2; Sek2; ARCC2; EC 2.7.10; EC 2.7.10.1; ECKepithelial cell receptor protein tyrosine kinase; EPH receptor A2; EphA2; ephrin type-A receptor 2; Epithelial cell kinase; soluble EPHA2 variant 1; Tyrosine-protein kinase receptor ECK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6680