Learn More
Biomatik Corporation Human JAK2 (Tyrosine-protein kinase JAK2) ELISA Kit
Supplier:  Biomatik Corporation EKF60668
Encompass_Preferred
Human JAK2 (Tyrosine-protein kinase JAK2) ELISA Kit for detecting JAK2 in cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O60674. Target Synonyms: JAK2; JAK-1; JAK1Atyrosine-protein kinase JAK1; JAK1B; Janus kinase 1; JTK3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6683