Learn More
Biomatik Corporation Human GPC1 (Glypican-1) ELISA Kit
Supplier:  Biomatik Corporation EKF60684
Encompass_Preferred
Human GPC1 (Glypican-1) ELISA Kit for detecting GPC1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P35052. Target Synonyms: GPC1; FLJ38078; glypican; glypican 1; glypican proteoglycan 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6696