Learn More
Biomatik Corporation Rat GROα/CXCL1 (Growth Regulated Oncogene Alpha) ELISA Kit
Supplier:  Biomatik Corporation EKF60691
Encompass_Preferred
Rat GROα/CXCL1 (Growth Regulated Oncogene Alpha) ELISA Kit for detecting GROα; CXCL1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.25-400pg/ml. Sensitivity: 3.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P14095. Target Synonyms: GROα; CXCL1; NAP3; GRO1; GRO-A; MGSA; MGSA-A; SCYB1; FSP; CINC-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6703