Learn More
Biomatik Corporation Mouse VEGFC (Vascular endothelial growth factor C) ELISA Kit
Supplier:  Biomatik Corporation EKF60693
Encompass_Preferred
Mouse VEGFC (Vascular endothelial growth factor C) ELISA Kit for detecting VEGFC in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P97953. Target Synonyms: VEGF-C; Vascular Endothelial cell Growth Factor C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6705