Learn More
Biomatik Corporation Chicken FSH (Follicle-Stimulating Hormone) ELISA Kit
Supplier:  Biomatik Corporation EKF60696
Encompass_Preferred
Chicken FSH (Follicle-Stimulating Hormone) ELISA Kit for detecting FSH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: Q8QGF8. Target Synonyms: FSH; Follicle-Stimulating Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6707