Learn More
Biomatik Corporation Porcine PAF (Platelet Activating Factor) ELISA Kit
Supplier:  Biomatik Corporation EKF60721
Encompass_Preferred
Porcine PAF (Platelet Activating Factor) ELISA Kit for detecting PAF in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: Platelet Activating Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6712