Learn More
Biomatik Corporation Human sCD163 (Soluble Cluster of Differentiation 163) ELISA Kit
Supplier:  Biomatik Corporation EKF60736
Encompass_Preferred
Human sCD163 (Soluble Cluster of Differentiation 163) ELISA Kit for detecting sCD163 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q86VB7. Target Synonyms: sCD163; Soluble Cluster of Differentiation 163; M130; MM130. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6717