Learn More
Biomatik Corporation Canine CKMB (Creatine Kinase MB Isoenzyme) ELISA Kit
Supplier:  Biomatik Corporation EKF60741
Encompass_Preferred
Canine CKMB (Creatine Kinase MB Isoenzyme) ELISA Kit for detecting CKMB in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P05124; P05123. Target Synonyms: CKMB; Creatine Kinase MB Isoenzyme; CK-MB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6719