Learn More
Biomatik Corporation Human CD147 (Extracellular Matrix Metalloproteinase Inducer) ELISA Kit
Supplier:  Biomatik Corporation EKF60749
Encompass_Preferred
Human CD147 (Extracellular Matrix Metalloproteinase Inducer) ELISA Kit for detecting CD147 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P35613. Target Synonyms: Emmprin; CD147; Extracellular Matrix Metalloproteinase Inducer; Basigin; BSG; 5F7; CD147 antigen; Collagenase stimulatory factor; EMMPRINTCSF; Extracellular matrix metalloproteinase inducer. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6723