Learn More
Biomatik Corporation Human MYL9 (Myosin regulatory light polypeptide 9) ELISA Kit
Supplier:  Biomatik Corporation EKF60760
Encompass_Preferred
Human MYL9 (Myosin regulatory light polypeptide 9) ELISA Kit for detecting MYL9 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P24844. Target Synonyms: MYL9; Myosin Light Chain 9; Regulatory; Myosin regulatory light polypeptide 9; Myosin regulatory light chain MRLC1; Myosin regulatory light chain 9; Myosin regulatory light chain 2; smooth muscle isoform; Myosin RLC; MLC-2C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6725