Learn More
Biomatik Corporation Canine CCL17 (C-C motif chemokine 17 ) ELISA Kit
Supplier:  Biomatik Corporation EKF60823
Encompass_Preferred
Canine CCL17 (C-C motif chemokine 17 ) ELISA Kit for detecting CCL17 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q95N01. Target Synonyms: C-C motif chemokine 17; CC chemokine TARC; Small-inducible cytokine A17; Thymus and activation-regulated chemokine; CCL17; TARC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6740