Learn More
Biomatik Corporation OCT2 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50003
Encompass_Preferred
OCT2 Colorimetric Cell-Based ELISA Kit for detecting 45201 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P09086. Gene ID: 5452. Target Synonyms: POU domain class 2 transcription factor 2; Lymphoid-restricted immunoglobulin octamer-binding protein NF-A2; Octamer-binding protein 2; Oct-2; Octamer-binding transcription factor 2; OTF-2; POU2F2; OCT2; OTF2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6756