Learn More
Biomatik Corporation Actin-γ2 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50016
Encompass_Preferred
Actin-γ2 Colorimetric Cell-Based ELISA Kit for detecting Actin-γ2 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P63267. Gene ID: 72. Target Synonyms: Actin gamma-enteric smooth muscle; Smooth muscle gamma-actin; Gamma-2-actin; Alpha-actin-3; ACTG2; ACTA3; ACTL3; ACTSG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6767