Learn More
Biomatik Corporation ATF3 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50044
Encompass_Preferred
ATF3 Colorimetric Cell-Based ELISA Kit for detecting ATF3 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P18847. Gene ID: 467. Target Synonyms: Cyclic AMP-dependent transcription factor ATF-3; cAMP-dependent transcription factor ATF-3; Activating transcription factor 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6792