Learn More
Biomatik Corporation BAD Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50053
Encompass_Preferred
BAD Colorimetric Cell-Based ELISA Kit for detecting BAD in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q92934. Gene ID: 572. Target Synonyms: BAD; BBC6; Bcl- XL/Bcl-2 associated death promoter; Bcl-2 binding component 6; Bcl-2-like 8 protein; Bcl-xL/Bcl-2 associated death promoter; Bcl2-antagonist of cell death; BCL2L8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6799