Learn More
Biomatik Corporation BIM Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50063
Encompass_Preferred
BIM Colorimetric Cell-Based ELISA Kit for detecting BIM in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O43521. Gene ID: 10018. Target Synonyms: BCL2 interacting mediator of cell death; BCL2-like protein 11; BCL2L11; BOD; Bcl-2 related ovarian death protein; Bcl2 interacting mediator of cell death; Bcl2-like protein 11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6806