Learn More
Biomatik Corporation Calsenilin/KCNIP3 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50079
Encompass_Preferred
Calsenilin/KCNIP3 Colorimetric Cell-Based ELISA Kit for detecting Calsenilin/KCNIP3 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9Y2W7. Gene ID: 30818. Target Synonyms: A-type potassium channel modulatory protein 3; CSEN; DRE-antagonist modulator; DREAM; KCNIP3; KChIP3; Kv channel-interacting protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6819