Learn More
Biomatik Corporation CD40 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50104
Encompass_Preferred
CD40 Colorimetric Cell-Based ELISA Kit for detecting CD40 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P25942. Gene ID: 958. Target Synonyms: B-cell surface antigen CD40; Bp50; CD40 antigen; CD40L receptor; CDw40; TNR5; tumor necrosis factor receptor superfamily member 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6836