Learn More
Biomatik Corporation Cyclin L1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50167
Encompass_Preferred
Cyclin L1 Colorimetric Cell-Based ELISA Kit for detecting Cyclin L1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9UK58. Gene ID: 57018. Target Synonyms: Cyclin-L1; Cyclin-L; CCNL1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6890