Learn More
Biomatik Corporation Human Histone H4 (HIST1H4A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC34050
Encompass_Preferred
The Human Histone H4 (HIST1H4A) ELISA Kit allows for quantitative determination of histone cluster 1, H4a in serum, plasma, tissue homogenates and cell lysates. Species Reactivity: Human. Detection Range: 37.5 pg/mL-2400 pg/mL. Sensitivity: 9.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P62805. Restriction: For Research Use Only.
Catalog No. 50-149-7971