Learn More
Biomatik Corporation FADD Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50215
Encompass_Preferred
FADD Colorimetric Cell-Based ELISA Kit for detecting FADD in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13158. Gene ID: 8772. Target Synonyms: FAS-associating death domain-containing protein; Mediator of receptor induced toxicity; MORT1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6934