Learn More
Biomatik Corporation FGFR3 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50225
Encompass_Preferred
FGFR3 Colorimetric Cell-Based ELISA Kit for detecting FGFR3 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P22607. Gene ID: 2261. Target Synonyms: FGFR-3; CD333 antigen; Fibroblast growth factor receptor 3; JTK4; CD333. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6943